Learn More
Thermo Scientific™ Orion™ AQUAfast™ Free & Total Chlorine Bulk DPD Powder Reagents
Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials are designed for high volume testing of residual chlorine in water samples.
Brand: Thermo Scientific™ AC71P1
Description
Measure residual chlorine in water samples using Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials with Orion AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. The powder chemistries dissolve quickly in the solution and are easy to mix. Each vial contains 250 counts of DPD reagent and is available for free chlorine or total chlorine analysis in the lab or field.
- Measure 0.02 to 2.00 mg/L (ppm) free chlorine or total chlorine
- Bulk powder dispensers are designed for high sample throughput – reduces the cost per test and minimizes waste compared with individually packaged reagents
- Equivalent to other widely used and recognized chemistries including Orion AQUAfast AC4P71 free chlorine and Orion AQUAfast AC4P72 total chlorine powder packs
- Compatible with Orion AQUAfast AQ4000, AQ3700 and AQ3170 colorimeters
- Compatible with Orion AquaMate AQ8000 and AQ7000 spectrophotometers
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
AQUAfast free chlorine DPD powder refill for AQ250F dispenser, 1 vial x 250 count | |
One free chlorine powder vial, 250 count | |
Powder | |
DPD |
AQUAfast AQ250F free chlorine DPD bulk powder dispenser | |
0.02 to 2 mg/L Cl2 | |
Chlorine |
Safety and Handling
- AQUAfast total chlorine powder pack
Signal Word
- Danger||Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P391-Collect spillage.
- P270-Do not eat, drink or smoke when using this product.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
Supplemental information
- MIXTURE LIST-Contains : sodium oxalate, N,N-diethylbenzene-1,4-diammonium sulphate