Learn More
Thermo Scientific Orion Orion Economy pH electrode gelled Ag/AgClelectrolyte, 0 to 12pH and 0° to 80°C, 1/Cs, MA:
Brand:  Thermo Scientific Orion 13-641-865A
Encompass
Orion Economy pH electrode gelled Ag/AgClelectrolyte, 0 to 12pH and 0° to 80°C, 1/Cs, MA:
Product Code. 12317259
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Thermo Scientific Orion Orion Economy pH electrode gelled Ag/AgClelectrolyte, 0 to 12pH and 0° to 80°C, 1/Cs, MA: