Learn More
Thermo Scientific™ Orion™ Combination pH Electrodes
Environmentally friendly lab pH electrodes to meet RoHS requirements by containing no lead, mercury or other hazardous substances — making disposal of old electrodes easy and hassle free.
Brand: Thermo Scientific™ GS9156BNWP
Description
- No disposal issues, no disposal cost
- Choice of single-junction models for clean-water measurements, or double-junction models for dirty-water testing
- Choice of refillable or low-maintenance gel-filled models
- Duarable epoxy body
- pH Range: 0 to 14
- Dimensions: 120 × 12mm, cap dia. 16mm
- Waterproof BNC connector, 1m cable; fit any meter with BNC input
- 12 months from date of purchase (GD9156BNWP)
- 6 months from date of purchase (GS9106BNWP and GD9106BNWP)
Services
- 12 months from date of purchase (GD9156BNWP)
- 6 months from date of purchase (GS9106BNWP and GD9106BNWP).

Specifications
Liquid-filled | |
Single | |
0 to 14 | |
Green Combination pH Electrode, liquid-filled | |
Orion pH Meter | |
1 m |
BNC (Waterproof) | |
Epoxy Body | |
0°C to 90°C | |
Liquid-filled | |
39.3 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.