Learn More
Thermo Scientific™ Orion™ Ion Selective Electrode Accessories
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
Brand: Thermo Scientific™ 948201
726.59 DKK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
Electrodes handles are used with plastic membrane combination or half-cell electrodes. Polishing strips help restore the performance of solid-state, half-cell, or combination ion selective electrodes.
Specifications
Polishing Strips for Solid State Ion Selective Electrodes | |
Ion Selective Electrode | |
Polishing Strips |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13