Learn More
Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes
Accurately measure ORP and temperature together in just one electrode, with a built-in temperature sensor.
2601.89 DKK - 4293.00 DKK
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | |||||
10691033
|
Thermo Scientific™
9179BN |
Epoxy Triode Low Maintenance | Orion A+ Series Meters |
2601.89 DKK
Each |
In Stock
Log in to see stock. |
|||||
11732857
|
Thermo Scientific™
9179BNMD |
Epoxy Triode Low Maintenance | Orion Star Series Meters |
2834.34 DKK
Each |
In Stock
Log in to see stock. |
|||||
10315173
|
Thermo Scientific™
9180BN |
Epoxy Refillable Triode | Orion A+ Series Meters | N/A | ||||||
10080514
|
Thermo Scientific™
9180BNMD |
Epoxy Refillable Triode | Orion Star Series Meters |
3486.70 DKK
Each |
In Stock
Log in to see stock. |
|||||
Description
Thermo Scientific™ Orion™ Triode 3-in-1 pH/Automatic Temperature Compensation Electrodes come with a built-in ATC and are engineered with an epoxy body for ruggedness. The 9179BN and 9179BNMD are low-maintenance gel-filled electrodes, best suited for educational institutes where minimal maintenance is required, while the 9180BNMD is a refillable electrode best suited for general purpose samples.
Orion™ Single Junction Electrodes provide a reliable option for routine ORP measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required
Electrode fill solution for 9180BNMD
900011 ORP filling solution for samples with total ionic strength > 0.2 M, 5 x 60 mL bottles
900001 ORP filling solution for samples with total ionic strength < 0.2 M, 5 x 60 mL bottles
Electrode storage solution
910001 4 M KCl storage solution, 475 mL bottle
Standards
967901 ORP standard, 475 mL bottle
967961 ORP standard, 5 x 60 mL bottles
pH cleaning solution kit
900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with 9810BNMD
900011 ORP filling solution for samples with total ionic strength > 0.2 M, 60mL 1 bottle
Warranty period
9179BN and 9179BNMD: 6 months from date of purchase
9180BNMD: 12 months from date of purchase
Specifications
Epoxy |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes