Learn More
Osmolality Standard, 2000mOsm/kg H2O, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
12 x 5 mL
Specifications
Specifications
Packaging | Ampoules |
Quantity | 12 x 5 mL |
Physical Form | Liquid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
OSM200023J1 | Certificate of Analysis | 26/03/2025 | RE-OSM-2000 |
OSM200022M1 | Certificate of Analysis | 09/04/2024 | RE-OSM-2000 |
OSM200022B1 | Certificate of Analysis | 08/03/2023 | RE-OSM-2000 |
OSM200021G1 | Certificate of Analysis | 05/01/2022 | RE-OSM-2000 |
OSM200021F1 | Certificate of Analysis | 02/11/2021 | RE-OSM-2000 |
Product Content Correction
Product Title
Osmolality Standard, 2000mOsm/kg H2O, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction