Learn More
Osmolality Standard, 850mOsm/kg H2O, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
12 x 5 mL
Specifications
Specifications
Packaging | Ampoules |
Quantity | 12 x 5 mL |
Physical Form | Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
OSM85024G1 | Certificate of Analysis | 25/02/2025 | RE-OSM-850 |
OSM85024C1 | Certificate of Analysis | 23/07/2024 | RE-OSM-850 |
OSM85023H1 | Certificate of Analysis | 22/04/2024 | RE-OSM-850 |
OSM85022K1 | Certificate of Analysis | 06/12/2023 | RE-OSM-850 |
OSM85022F1 | Certificate of Analysis | 06/12/2022 | RE-OSM-850 |
Product Content Correction
Product Title
Osmolality Standard, 850mOsm/kg H2O, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction