Learn More
Kimberly-Clark™ KleenGuard™ A40 Light Duty Overshoe with Sole
Ideal for keeping footwear clean and covered. KleenGuard™ A40 Light Duty Overshoe with Sole reduces contamination in food processing and light industrial areas.
5625.00 DKK
Specifications
Color | White |
---|---|
Material | Spundbond Polypropylene |
Toe Material | Multi-layer Non-woven Barrier Laminate |
Waterproof | Water Resistant |
Product Type | Overshoe |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
12685576
|
Kimberly-Clark™
98710 |
Medium/Large |
5625.00 DKK
Pack of 200 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
12695576
|
Kimberly-Clark™
98720 |
X-Large/2X-Large |
5625.00 DKK
Pack of 200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- Elasticated overshoe with sole
- Single-use light duty
- Ideal for keeping footwear clean and covered
- Reduce contamination in food processing and light industrial areas
Specifications
White | |
Multi-layer Non-woven Barrier Laminate | |
Overshoe |
Spundbond Polypropylene | |
Water Resistant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Kimberly-Clark™ KleenGuard™ A40 Light Duty Overshoe with Sole