Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs.
Brand: Thermo Scientific™ D1-10D
This item is not returnable.
View return policy
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with Owl™ D2 Wide-Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Specifications
Owl™ D2 Wide Gel Electrophoresis System | |
Standard Comb | |
1.5 mm |
Owl | |
10 |
Owl D2 Wide Gel System Illustration
Version: FEB.2016
File size: 78.5 kB
Owl D2 Gradient Gel Caster Instructions
Version: FEB.2016
File size: 137.5 kB
Product Content Correction
Product Title
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs > Standard; 10 teeth; 1.5mm thick
Spot an opportunity for improvement?Share a Content Correction