missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
CAS | 144-62-7 |
Molecular Formula | C2H2O4 |
InChI Key | MUBZPKHOEPUJKR-UHFFFAOYSA-N |
SMILES | OC(=O)C(O)=O |
IUPAC Name | oxalic acid |
Molecular Weight (g/mol) | 90.03 |
Quantity | 1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Oxalic acid, 0.05 M (0.1 N), Scharlab >
Spot an opportunity for improvement?Share a Content Correction