missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ Oxivir Excel Disinfectant Wipes
Pre-moistened cleaner disinfectant wipes for non-invasive medical devices and nonporous surfaces
Brand: Diversey™ 101105022
This item is not returnable.
View return policy
Description
- One-step cleaner disinfectant with broad spectrum efficacy and short contact times
- Oxivir Excel Wipe can be used on non-invasive medical devices and nonporous surfaces
- Full virucidal claim including non-enveloped and enveloped viruses such as SARS-CoV-2 in just 30 seconds
- Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
- No hazard classification
- Powered by AHP Technology (Accelerated Hydrogen Peroxide)
- AHP formulation is gentle on surfaces extending equipment life
- Excellent user safety profile - no PPE required
- Does not contain respiratory irritants
- Suitable for use in high risk areas including ICU, OR, NICU and patient rooms
- Fragrance free
- No rinsing required
- Safer for the environment and user, since the AHP actives break down into water and oxygen. Free from Volatile Organic Compounds (VOCs)
- Medical Device Class IIa
Specifications
All Purpose Wipes | |
15 cm | |
17 cm | |
For DK, FI, GB, NO, and SE only | |
Canister, PT02 | |
160 Wipes/Pk., 6 Pk. | |
Efficacy proven by compliance with EN13727, EN13624, EN14476 and EN16615 |
6 in. | |
7 in. | |
Polypropylene | |
15 x 17 cm | |
All purpose virucidal wipes | |
Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Diversey™ Oxivir Excel Disinfectant Wipes > Dimensions (L x W): 15 x 17 cm (6 x 7 in.); Format: Canister, PT02; Item Description: For DK, FI, GB, NO, and SE only; Packaging Quantity: 160 Wipes/Pk, 6 Pk
The surfactants used in this product are biodegradable in compliance with the requirement of EU Detergent Regulation, EC 648/2004
Spot an opportunity for improvement?Share a Content Correction