Learn More
Thermo Scientific™ Oxoid™ AnaeroGen™ Compact
Oxoid AnaeroGen Compact is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
Brand:  Thermo Scientific™ AN0010C
430.03 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10729081
  • 492.00 DKK / Pack of 10
Edge
Explore more special offers
Description
Description
Oxoid AnaeroGen Compact is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
  • Use 1 sachet / Compact Plastic Pouch
  • Use with AnaeroGen™ Compact Sealing Clip, Part No. AN0005C
Includes: 10 sachets and 10 pouches.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AnaeroGen Compact
10 sachets and 10 pouches
Thermo Scientific Oxoid
10/Pk.
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6251120 Certificate TypeCertificate of Analysis Date13/04/2025 Catalog NumberAN0010C
Lot Number6245554 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog NumberAN0010C
Lot Number6244628 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog NumberAN0010C
Lot Number6241829 Certificate TypeCertificate of Analysis Date08/03/2025 Catalog NumberAN0010C
Lot Number6238130 Certificate TypeCertificate of Analysis Date08/03/2025 Catalog NumberAN0010C
Product Title
Thermo Scientific™ Oxoid™ AnaeroGen™ Compact >