Learn More
Thermo Scientific™ Oxoid™ AnaeroGen™ W-Zip Compact Gas Generator System
Oxoid AnaeroGen W-Zip Compact is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
Brand:  Thermo Scientific™ AN0010W
593.61 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12201972
  • 728.00 DKK / Pack of 10
Description
Oxoid AnaeroGen W-Zip Compact is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
  • Use 1 sachet / Compact Plastic Pouch
Includes: 10 sachets and 10 W-Zip pouches.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AnaeroGen W-Zip Compact
10 sachets and 10 W-Zip pouches
10 Each/Pk.
Compact Gas Generator Kit
Thermo Scientific Oxoid
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6265487 Certificate TypeCertificate of Analysis Date09/05/2025 Catalog NumberAN0010W
Lot Number6254194 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog NumberAN0010W
Lot Number6226843 Certificate TypeCertificate of Analysis Date01/02/2025 Catalog NumberAN0010W
Lot Number6208790 Certificate TypeCertificate of Analysis Date21/12/2024 Catalog NumberAN0010W
Lot Number6208732 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog NumberAN0010W
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ AnaeroGen™ W-Zip Compact Gas Generator System > Pack of 10