Learn More
Thermo Scientific™ EE Broth (Buffered Glucose Brilliant Green Bile Broth) (Dehydrated)
Enrich Enterobacteriaceae in bacteriological examination of foods and animal feed stuffs with this selective medium.
1535.00 DKK
Specifications
Description | EE Broth |
---|---|
Form | Powder |
Product Type | Dehydrated Culture Media |
Description
Enrich Enterobacteriaceae in bacteriological examination of foods1 and animal feed2 stuffs with Thermo Scientific™ Oxoid™ EE Broth (Buffered Glucose Brilliant Green Bile Broth) (Dehydrated). This medium is more inhibitory to non-Enterobacteriaceae than other non-selective media, e.g. Mannitol broth3 or Lactose broth4 by virtue of the presence of brilliant green and bile salts in the preparation.
To monitor the sanitary quality of food and drugs, it is important to enumerate Enterobacteriaceae in samples accurately. E.E. Broth is recommended as an enrichment medium for Enterobacteriaceae in the bacteriological examination of foods and animal feedstuffs.
- Selective: non-Enterobacteriaceae are inhibited due to brilliant green and bile salts in the medium
- Reliable: overcomes inhibition of small numbers of Enterobacteriaceae cells due to bile salt variations
- Accurate: the entire Enterobacteriaceae group can be used as indicator organisms avoiding discrepancies which can occur with standard `coli-aerogenes’ test
The medium is more inhibitory to non-Enterobacteriaceae than other non-selective media (e.g., Mannitol broth or Lactose broth) due to the presence of brilliant green and bile salts.
The reliability of methods used to enumerate Enterobacteriaceae depends on resuscitation of damaged cells. Weakened cells arise from exposure to dehydration, low pH and other unfavorable conditions. E.E. Broth’s formulation includes purified ox bile to overcome inhibition of small numbers of Enterobacteriaceae cells due to bile salt variations. A preliminary assay can be used to check growth by inoculating approximately one viable cell per medium unit.
For the bacteriological evaluation of processed foods, the entire Enterobacteriaceae group can be used as indicator organisms. This overcomes discrepancies that can arise when lactose-negative, anaerogenic lactose-positive or late lactose fermenting enterobacteria are present but are missed by the standard `coli-aerogenes’ tests.
To overcome these problems, lactose media have been replaced by those containing glucose. Mossel et al1 cited several examples which referred to foods contaminated with salmonellae, although results for coliforms were negative. A later example quoted by Mossel2 involved an outbreak of diarrhea caused by French mold-fermented soft cheese contaminated by Escherichia coli serotype O124. This organism was lactose-negative and, therefore, was not detected in coliform tests, but only recognized when the commodity was tested for Enterobacteriaceae since it fermented glucose rapidly.
E.E. Broth should be used as an enrichment broth in conjunction with Violet Red Bile Glucose Agar (Part No. PO5043A). When specific organisms are required, rather than Enterobacteriaceae in general, subcultures must be made onto lactose differential media, e.g., Desoxycholate Citrate Agar.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
EE Broth | |
Powder | |
Dehydrated Culture Media |
Safety and Handling
missing translation for 'shelfLife' : 1 year
For Laboratory Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.