Learn More
Thermo Scientific™ Oxoid™ Egg Yolk Tellurite Emulsion
Brand: Thermo Scientific™ SR0054C
489.97 DKK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
Description
A sterile emulsion of egg yolk containing potassium tellurite for use in Baird-Parker Medium (CM0275).
Specifications
100 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6251116 | Certificate of Analysis | 25/04/2025 | SR0054C |
6254207 | Certificate of Analysis | 25/04/2025 | SR0054C |
6235812 | Certificate of Analysis | 14/04/2025 | SR0054C |
6244684 | Certificate of Analysis | 08/04/2025 | SR0054C |
6238239 | Certificate of Analysis | 28/03/2025 | SR0054C |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30