Learn More
Thermo Scientific™ Oxoid™ IMAGEN™ Respiratory Screen Positive and Negative Control Slide
Slide with 1 row of wells containing separate preparations from cell cultures infected with RSV, influenza A and B virus, parainfluenza virus 1, 2 and 3 or adenovirus.
Brand:  Thermo Scientific™ S612130-2
1386.39 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 13205329
  • 1410.00 DKK / Each
Description
The IMAGEN™ Control Slides are for use as a control to the following tests: IMAGEN Adenovirus (K610011-2) IMAGEN Respiratory Syncyti al Virus (RSV) (K610211-2) IMAGEN Parainfluenza Group (K610311-2) IMAGEN Parainfluenza Typing 1, 2 and 3 (K610411-2) IMAGEN Influenza A and B Virus (K610511-2) IMAGEN Respiratory Screen (K612011-2) IMAGEN hMPV (K612511-2) IMAGEN Herpes Simplex Virus (HSV) (K610611-2) IMAGEN Chlamydia kit (K610111-2)
The second row of wells contains preparations from uninfected cell cultures for use as negative controls. Sealed in foil pouch filled with nitrogen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IMAGEN™ Respiratory Screen Positive and Negative Control Slide
Control
1 Slide/Pk.
Documents
Instructions
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6210498 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberS612130-2
Lot Number542388 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog NumberS612130-2
Lot Number3581515 Certificate TypeCertificate of Analysis Date09/03/2023 Catalog NumberS612130-2
Lot Number3350943 Certificate TypeCertificate of Analysis Date08/10/2021 Catalog NumberS612130-2
Lot Number3214555 Certificate TypeCertificate of Analysis Date29/01/2021 Catalog NumberS612130-2
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ IMAGEN™ Respiratory Screen Positive and Negative Control Slide > 1 Slide/Pk.