Learn More
Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Supplement
Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Supplement is for the selective enrichment of Salmonella spp. from food samples in 24 hours in a single enrichment step.
Brand: Thermo Scientific™ SR0242E
287.52 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Supplement is for the selective enrichment of Salmonella spp. from food samples in 24 hours in a single enrichment step.
- Add to ONE Broth-Salmonella Part No. CM1091B
- Each vial supplements 500mL of medium

Specifications
Salmonella species | |
Freeze-dried | |
10/Pk. |
ONE Broth-Salmonella Supplement | |
Microbiology Media | |
Each vial for 225mL medium |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6169127 | Certificate of Analysis | 16/09/2024 | SR0242E |
3793400 | Certificate of Analysis | 28/02/2024 | SR0242E |
3718392 | Certificate of Analysis | 14/09/2023 | SR0242E |
3639482 | Certificate of Analysis | 06/04/2023 | SR0242E |
3563298 | Certificate of Analysis | 24/10/2022 | SR0242E |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.