Learn More
Thermo Scientific™ Oxoid™ Prepared Sabouraud Dextrose Agar Bottled Media
Brand:  Thermo Scientific™ BO0408T
1225.50 DKK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Product Code. 12647228
  • 1325.00 DKK / Pack of 10
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4506123 Certificate TypeCertificate of Analysis Date11/05/2025 Catalog NumberBO0408T
Lot Number4498275 Certificate TypeCertificate of Analysis Date18/03/2025 Catalog NumberBO0408T
Lot Number4493965 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberBO0408T
Lot Number4487241 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberBO0408T
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Prepared Sabouraud Dextrose Agar Bottled Media > Volume: 250mL