Learn More
Thermo Scientific™ Oxoid™ Replacement Desiccant, Dispenser
Maintain a low moisture environment when storing AST products used for manual Antimicrobial Susceptibility Testing.
Brand:  Thermo Scientific™ ST9100
280.57 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10200343
  • 330.00 DKK / Each
Description
Maintain a low moisture environment when storing AST products for manual Antimicrobial Susceptibility Testing with Thermo Scientific™ Oxoid™ Replacement Desiccant (Dispenser).
The Oxoid Replacement Desiccant fits both the Oxoid 8- and 12-position antimicrobial susceptibility disc dispensers.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Oxoid Replacement Desiccant (Dispenser)
Replacement Desiccant
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6272669 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberST9100
Lot Number6263409 Certificate TypeCertificate of Analysis Date29/04/2025 Catalog NumberST9100
Lot Number6235784 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog NumberST9100
Lot Number6235782 Certificate TypeCertificate of Analysis Date13/02/2025 Catalog NumberST9100
Lot Number6223633 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberST9100
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Replacement Desiccant, Dispenser >