Learn More
Thermo Scientific™ Oxoid™ V Factor Discs
Thermo Scientific™ Oxoid V Factor Disc is used for identification of Haemophilus spp.
Brand: Thermo Scientific™ DD0004T
193.03 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Oxoid V Factor Disc is used for identification of Haemophilus spp.
Ordering Alerts:
50 Discs/Cartridge
Specifications
-10°C to -20°C | |
Haemophilus species | |
1 Cart./Pk. |
V Factor Discs | |
X Factor Disc | |
50 Disks/Cartridge |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6172450 | Certificate of Analysis | 16/10/2024 | DD0004T |
3672810 | Certificate of Analysis | 06/01/2024 | DD0004T |
3721997 | Certificate of Analysis | 06/01/2024 | DD0004T |
3672810 | Certificate of Analysis | 01/08/2023 | DD0004T |
3531404 | Certificate of Analysis | 01/02/2023 | DD0004T |