Learn More
Thermo Scientific™ Oxoid™ Vancomycin Supplement
Used for addition to selective media for isolation of Vancomycin Resistant Enterococci and High Level Aminoglycoside Resistant Enterococci from clinical specimens.
Brand: Thermo Scientific™ SR0186E
694.42 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Oxoid Vancomycin Supplement is for addition to selective media for the isolation of Vancomycin Resistant Enterococci (VRE) and High Level Aminoglycoside Resistant Enterococci (HLARE) from clinical specimens.
- Each vial supplements 500mL of medium
Specifications
Oxoid™ Vancomycin Supplement | |
Freeze-dried | |
Microbiology Media | |
10/Pk. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6180778 | Certificate of Analysis | 04/11/2024 | SR0186E |
6101249 | Certificate of Analysis | 09/05/2024 | SR0186E |
3725299 | Certificate of Analysis | 28/09/2023 | SR0186E |
3618210 | Certificate of Analysis | 21/02/2023 | SR0186E |
3527882 | Certificate of Analysis | 08/12/2022 | SR0186E |