missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OxPhos Complex IV subunit IV Monoclonal Antibody (10G8D12C12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A21347
This item is not returnable.
View return policy
Description
OxPhos Complex IV subunit IV Monoclonal antibody specifically detects OxPhos Complex IV subunit IV in Bovine, Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
OxPhos Complex IV subunit IV | |
Monoclonal | |
1 mg/mL | |
HEPES buffered saline with 0.02% sodium azide | |
Mouse | |
purified | |
RUO | |
Bovine, Human | |
Antibody | |
IgG2a κ |
Immunohistochemistry (Paraffin), Western Blot | |
10G8D12C12 | |
Unconjugated | |
COX4; COX4-1; COXIV; COX IV-1; cytochrome c oxidase polypeptide IV; cytochrome c oxidase subunit 4 isoform 1 mitochondrial | |
Bovine Complex IV subunit IV | |
100 μg | |
Primary | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction