Learn More
P-Cadherin Monoclonal Antibody (CSTEM29), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-2237-95
Description
This monoclonal antibody CSTEM29 reacts with human P-Cadherin, also known as Cadherin-3. Applications Reported: This CSTEM29 antibody has been reported for use in flow cytometric analysis and immunoblot. Applications Tested: This CSTEM29 antibody has been tested by flow cytometric analysis of normal human induced pluripotent stem cells, and for western blot. For flow cytometry, this may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. For western blot, this can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
P-Cadherin Monoclonal antibody specifically detects P-Cadherin in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western BlotSpecifications
P-Cadherin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P10287, P22223 | |
Mouse | |
10 mg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
CSTEM29 | |
Unconjugated | |
Cdh3 | |
AI385538; CADH3; cadherin 15, M-cadherin (myotubule); cadherin 3; cadherin 3, type 1, P-cadherin (placental); cadherin-3; Cadp; calcium-dependent adhesion protein, placental; CDH15; CDH3; CDHP; HJMD; PCAD; P-cadherin; Placental cadherin; Placental-cadherin; RPE-specific cadherin | |
Affinity chromatography | |
RUO | |
1001, 12560 | |
4° C, do not freeze | |
Liquid |