missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ p16/CEN9q FISH Probe

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

12030.00 DKK - 19810.00 DKK

Specifications

Content And Storage Store at 4°C in the dark.
Label or Dye FITC, Texas Red
For Use With (Application) Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
Includes DAPI Counterstain (1500 ng/mL ) 250 uL
Location 9p21, 9q21
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16022975
View Documents
Abnova™
FG0002.200uL
200 μL
19810.00 DKK
200µL
Please to purchase this item. Need a web account? Register with us today!
16012975
View Documents
Abnova™
FG0002.100uL
100 μL
12030.00 DKK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.

Advantages

  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity

Specifications

Specifications

Store at 4°C in the dark.
Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
9p21, 9q21
p16/CEN9q FISH Probe
FITC, Texas Red
DAPI Counterstain (1500 ng/mL ) 250 uL
p16, CEN9q
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.