missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Rabbit anti-Human, Rat, Clone: 9L6E2, Novus Biologicals™
Rabbit Monoclonal Antibody
1460.01 DKK - 3692.13 DKK
Specifications
Antigen | p53 |
---|---|
Clone | 9L6E2 |
Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18304835
|
Bio-Techne
NBP3-15792-20UL |
20 μg |
1460.01 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18310803
|
Bio-Techne
NBP3-15792-100UL |
100 μg |
3692.13 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p53 Monoclonal antibody specifically detects p53 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDownSpecifications
p53 | |
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-393 of human p53 (NP_000537.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
9L6E2 | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7157 | |
IgG | |
Affinity purified |