missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pan Coronavirus Spike Antibody (38), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP3-14660AF350
This item is not returnable.
View return policy
Description
Pan Coronavirus Spike Monoclonal antibody specifically detects Pan Coronavirus Spike in Virus samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Pan Coronavirus Spike | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
38 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Antibody | |
IgG1 |