missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pan Nuclear Pore Complex Marker Antibody (39C7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP1-05390
This item is not returnable.
View return policy
Description
pan Nuclear Pore Complex Marker Monoclonal specifically detects pan Nuclear Pore Complex Marker in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Drosophila, Equine, Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
pan Nuclear Pore Complex Marker | |
Monoclonal | |
Unconjugated | |
Supplied as concentrated hybridoma cell culture media. with 5mM Sodium Azide | |
Mouse | |
62 kDa | |
0.5 mL | |
Core ESC Like Genes, Stem Cell Markers | |
Panspecific | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
39C7 | |
Western Blot 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:500 | |
107 kDa nucleoporin, NPC proteins, Nuclear pore complex protein Nup107, Nucleoporin 107kDa, Nucleoporin Nup107, NUP 107, NUP 84, NUP107, NUP84 | |
Yeast nuclear preparations | |
Tissue culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, C. elegans, Chicken, Drosophila, Equine, Yeast | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
pan Nuclear Pore Complex Marker Antibody (39C7), Novus Biologicals™ > 0.5 mL; Unconjugated
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction