Learn More
Novus Biologicals™ PAP Pen Fine Tip
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Specifications
Specifications
Content And Storage | Room Temperature |
Quantity | 1 Each |
For Use With (Application) | Preparation of Working Solutions 1. Ready to be Used Directly on Wet Slides. Protocol Recommendations 1. Hold The Pen Point up and Press it with Your Fingertip to Take Blocked Air Out From The Pen Barrel to Prevent Overflow. 2. To Start, Press The Tip of The Pen Straight Down Until Fluid Starts Soaking The Tip and Then Draw a Circle Around The Specimen. |
Type | Tools and Accessories |
Product Content Correction
Product Title
Novus Biologicals™ PAP Pen Fine Tip >
Spot an opportunity for improvement?Share a Content Correction