Learn More
Brand™ PARAFILM™ M Cutter
Provides convenient clean storage and cutting of PARAFILM™ M Sealing Film. BRAND™ PARAFILM™ M Cutter accommodates 50mm and 100mm wide rolls.
Brand:  Brand™ 701650
1426.16 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10578491
  • 2130.00 DKK / Each
Only null left
Estimated Shipment: 11-08-2025
Description
The handy PARAFILM™ M Cutter is convenient for clean storage and cutting of PARAFILM™ M sealing film. Suitable for 50 mm and 100 mm wide rolls. Easy to use: 1. Inserting, 2. Dispensing, 3. Cutting, 4. Taking.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FIlm cutter
50 and 100 mm W PARAFILM M Sealing film
1 Ea.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Brand™ PARAFILM™ M Cutter > For 50mm and 100mm Wide Rolls