Learn More
Cole-Parmer™ Parker Hannifin™ PFA Compression Female Threaded Adapter
Excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Brand:  Cole-Parmer™ 31320-72
 View more versions of this product
Product Code. 11769729
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Superior chemical resistance with durable PFA composition.
  • Get a tight seal at higher pressures.
  • Withstand higher pressures than barbed fittings.
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
Specifications
Compression x NPT(F)
PFA
1/4 x 1/2 in.
1/4 in. NPT(F)
Compression Threaded Adapter
Autoclavable
1/2 in.
Autoclave or Ethylene Oxide
Clear
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Parker Hannifin™ PFA Compression Female Threaded Adapter > 1/2 in. tubing OD x 1/4 in. NPT(F)