Learn More
Cole-Parmer™ Parker Hannifin™ PFA Compression Female Threaded Adapter
Excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Brand: Cole-Parmer™ 31320-52
This item is not returnable.
View return policy
Description
- Superior chemical resistance with durable PFA composition.
- Get a tight seal at higher pressures.
- Withstand higher pressures than barbed fittings.
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
Compression x NPT(F) | |
PFA | |
1/8 x 1/4 in. | |
1/8 in. NPT(F) | |
Compression Threaded Adapter |
Autoclavable | |
1/4 in. | |
Autoclave or Ethylene Oxide | |
Clear |
Product Content Correction
Product Title
Cole-Parmer™ Parker Hannifin™ PFA Compression Female Threaded Adapter > 1/4 in. tubing OD x 1/8 in. NPT(F)
Spot an opportunity for improvement?Share a Content Correction