Learn More
Mettler Toledo™ Parking Beaker Insert, EasyPlace
The parking beaker insert should be used while using the parking beaker with storage solution.
Brand:  Mettler Toledo™ 30671572
55.96 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17561016
  • 57.40 DKK / Each
Focus On Clean Water
Use promo code 24139 at checkout
View Available Promotions
Only null left
In Stock
Description
  • Safe Short-Term Storage
  • The parking beaker insert holds storage solutions and can be placed in the parking beaker
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Parking Beaker Insert, EasyPlace
EasyPlace
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Shop now for up to 60% off Clean Water Research essentials
    Promo Code: 24139
    Expires2025-08-21
    New Offer
    Save Now
Product Content Correction
Product Title
Mettler Toledo™ Parking Beaker Insert, EasyPlace > Parking Beaker Insert