missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Paroxetine maleate
Highly potent and selective 5-HT uptake inhibitor
1605.00 DKK - 5530.00 DKK
Specifications
Purity | >99% |
---|---|
Product Type | Paroxetine maleate |
Molecular Weight (g/mol) | 445.44 |
Inhibitors | Paroxetine maleate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18083782
|
Tocris Bioscience™
2141/10 |
10 mg |
1605.00 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18798566
|
Tocris Bioscience™
2141/50 |
50 mg |
5530.00 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Highly potent and selective 5-HT uptake inhibitor that binds with high affinity to the serotonin transporter (Ki = 0.05 nM). Ki values are 1.1, 350 and 1100 nM for inhibition of [3H]-5-HT, [3H]-l-NA and [3H]-DA uptake respectively. Displays minimal affinity for α1-, α2- or β-adrenoceptors, 5-HT2A, 5-HT1A, D2 or H1 receptors at concentrations below 1000 nM, however displays weak affinity for muscarinic ACh receptors (Ki = 42 nM). Antidepressant and anxiolytic in vivo.Specifications
>99% | |
Paroxetine maleate | |
445.44 | |
Paroxetine maleate |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ Paroxetine maleate