missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Pasteur-Plast 2.5 mL Graduated Pipettes

Pasteur-Plast Pipettes render liquid handling simple and safe with an unbreakable design.

Brand:  Ratiolab™ 2600131

Product Code. 10051128

  • 261.00 DKK / Pack of 500
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Cross-contamination and carry-over eliminated so that tedious clean-ups are unnecessary
  • No danger of glass breakage with resulting danger of infectious material escaping
  • Made of transparent polyethylene
  • Inert to biological fluids and most acids
  • Low affinity of the surface reduces the loss of cells and valuable proteins through binding
  • Soft, elastic bulb
  • Burr-free orifice for neat, clean and precise dispensing
  • With clearly visible embossed graduations
  • BSE-free
  • Food-safe
  • Minimal residual wetting due to the controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Provided in packs of 500
Specifications

Specifications

Polyethylene
Embossed
2.5 mL
150 mm
Yes
500/Pk.
Transparent
Pipette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ratiolab™ Pasteur-Plast 2.5 mL Graduated Pipettes > 2.5 mL; 150 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.