missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ratiolab™ Pasteur-Plast Pipets
Render liquid handling simple and safe with these Pasteur-Plast pipets, which are inert to biological fluids and most acids. The low affinity of the surface reduces the loss of cells and valuable proteins through binding.
Brand: ratiolab™ 2600190
This item is not returnable.
View return policy
Description
- Soft, elastic bulb is unbreakable, thus reducing the risk of glass injury
- Burr-free orifice for neat, clean and precise dispensing
- Only externally graduated Pasteur-plast pipets with clearly visible embossed graduations
Specifications
LDPE | |
No | |
≤70°C | |
10 mL | |
Pasteur Pipet |
Non-sterile | |
200 Pipets/Box | |
Clear | |
196 mm | |
Single-use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ratiolab™ Pasteur-Plast Pipets > Pasteur-Plast Pipets; Graduated: No; Length (Metric): 196 mm; Sterility: Non-sterile; Volume (Metric): 10 mL
Spot an opportunity for improvement?Share a Content Correction