Learn More
PBS, 10X Solution, Molecular Biology Grade Product Code.: 15825418
PBS, 10X Solution, Molecular Biology Grade
Product Code. 15825418 Shop All Thermo Scientific Products
1 L, 1 L
Click to view available options
Quantity:
100 mL
1 L
5 L
Unit Size:
1 L
100mL
5 L
Product Code. 15825418
Brand: Thermo Scientific J75889.K2
Request Bulk or Custom Format
Edge
For biological research. Affymetrix/USB™ PBS, 10X Solution, Molecular Biology Grade is 0.2μm filtered and dispensed into durable, square nalgene bottles and upright carton.
  • Consists of 80.6mM sodium phosphate, 19.4mM potassium phosphate, 27mM KCl and 1.37M NaCl in high purity dH2O
  • Filtered and dispensed into durable, square Nalgene bottles and upright carton with convenient dispensing spout
  • Dilute to 1X with high purity dH2O for a solution consisting of 10mM phosphate
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 L
Formulation 80.6 mM sodium phosphate, 19.4 mM potassium phosphate, 27 mM KCl and 1.37 M NaCl in high purity dH2O
pH 7.4
Form Solution
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number241356 Certificate TypeCertificate of Analysis Date23/07/2024 Product CodeJ75889.AE,J75889.K2,J75889.K8
Lot Number234377 Certificate TypeCertificate of Analysis Date09/07/2024 Product CodeJ75889.AE,J75889.K2,J75889.K8
Lot Number202110 Certificate TypeCertificate of Analysis Date13/11/2023 Product CodeJ75889.AE,J75889.K2,J75889.K8
Lot Number204161 Certificate TypeCertificate of Analysis Date13/11/2023 Product CodeJ75889.AE,J75889.K2,J75889.K8
Lot Number231845 Certificate TypeCertificate of Analysis Date08/09/2023 Product CodeJ75889.AE,J75889.K2,J75889.K8
RUO – Research Use Only
Product Content Correction
Product Title
PBS, 10X Solution, Molecular Biology Grade >