missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific PC Tube, 100 mL
100 mL PC Tube.
Brand: Thermo Scientific 76009094
This item is not returnable.
View return policy
Description
- 100 mL PC Tube
Specifications
100 mL | |
Polycarbonate | |
3.4 oz. | |
Centrifuge Tube |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction