missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCP4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1250.51 DKK
Specifications
Antigen | PCP4 |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:100-1:500 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PCP4 Polyclonal antibody specifically detects PCP4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
PCP4 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
brain specific polypeptide PEP19, 10Brain-specific antigen PCP-4, Brain-specific polypeptide PEP-19, PEP19, PEP-19, Purkinje cell protein 4 | |
Produced in rabbits immunized with E. coli-derived Human PCP4 fragment, | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry-Paraffin 1:100-1:500 | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.0 with 0.03% Proclin 300 | |
5121 | |
IgG | |
Antigen and protein A Affinity-purified |