Learn More
BRAND™ 8-Strip PCR Caps, BIO-CERT™ PCR QUALITY
BRAND™ PCR cap strips ensure reliable closures for PCR strips and plates, safeguarding your samples against evaporation and contamination.
Brand: BRAND™ 781343
241.73 DKK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
This item is not returnable.
View return policy
Description
- Exceptional Quality and Purity: BIO-CERT™ PCR QUALITY ensures caps are free from human DNA, RNase, DNase, pyrogens, and PCR inhibitors.
- Secure Sealing: Designed to reliably seal PCR strips and plates with 24-, 48-, or 96-wells, effectively protecting your samples from evaporation and contamination.
- Perfect Compatibility: Optimally shaped to match BRAND™ PCR tube strips and plates, ensuring easy opening and closing. They are also compatible with most other manufacturers, offering flexibility for your workflow.
- Contamination Prevention: Equipped with side tabs for precise positioning and a small opening attachment on each cap to prevent direct contact with the interior.
- qPCR-Ready Transparency: Highly transparent flat caps enable accurate fluorescence measurement, making them ideal for qPCR applications.
- Enhanced Sealing with Domed Caps: Domed caps provide a stronger sealing effect when used with thermal cyclers lacking pressure sensors.
Specifications
Green | |
Domed | |
Polypropylene | |
Yes | |
Autoclavable | |
Single-use |
Microplates | |
Non-sterile | |
Yes | |
8-strip Domed Cap | |
Yes | |
Cap Strip |
Brand PCR Products
File size: 7.8 MB
Technical Note: PCR sealing options
File size: 6.4 MB
Product Content Correction
Product Title
BRAND™ 8-Strip PCR Caps, BIO-CERT™ PCR QUALITY > Color: Green; Cap Type: Domed
Spot an opportunity for improvement?Share a Content Correction