Learn More
Thermo Scientific™ PCR Plate, 96-well, low profile, skirted, Sustain™ Series
The Thermo Scientific PCR Plate, 96-well, low profile, skirted, Sustain Series, has a low-profile, thin well wall design and is compatible with PCR and qPCR applications.
Brand:  Thermo Scientific™ AB0800SS
Product Code. 30179690
  • 972.00 DKK / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Show More
Specifications
Specifications
Custom Barcoding Available
SBS Standard
Natural
Polypropylene
Less waste, renewable material
Certified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free
Autoclavable
Skirted
96
25 plates
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ PCR Plate, 96-well, low profile, skirted, Sustain™ Series > 25 plates