Learn More
Tocris Bioscience™ PD 98059
MEK inhibitor
Brand: Tocris Bioscience™ 1213/50
5082.70 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
PD 98059 is an inhibitor of mitogen-activated protein kinase kinase (MKK/ MEK). Acts by binding to the inactivated form of MEK, thereby preventing its phosphorylation by cRAF or MEK kinase (IC50 = 2-7 μM). Inhibits cell growth and proliferation in acute myelogenous leukemia (AML) cell lines; causes G1 arrest by blocking p53-dependent p21 induction. Enhances embryonic stem cell self-renewal.Specifications
98% | |
167869-21-8 | |
50 mg | |
M.W. (Observed): 267.28 |
2-(2-Amino-3-methoxyphenyl)-4 H-1-benzopyran-4-one | |
Room Temperature | |
PD 98059 | |
PD 98059 |
For Research Use Only