missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals PDE1B Antibody (4F9), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals H00005153-M02
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PDE1B Monoclonal antibody specifically detects PDE1B in Human samples. It is validated for Western Blot, ELISASpezifikation
PDE1B | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
63 kDa Cam-PDE, calcium/calmodulin-dependent 3'-5'-cyclic nucleotide phosphodiesterase 1B, Cam-PDE 1B, EC 3.1.4, EC 3.1.4.17, PDE1B1calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, PDES1Bcalmodulin-stimulated phosphodiesterase PDE1B1, phosphodiesterase 1B, calmodulin-dependent, phosphodiesterase IB, calmodulin-dependent, Phosphodiesterase-1B, presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase familyknown as PDE1B | |
PDE1B (AAH32226, 437 a.a. ~ 536 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TDVAEKSVQPLADEDSKSKNQPSFQWRQPSLDVEVGDPNPDVVSFRSTWVKRIQENKQKWKERAASGITNQMSIDELSPCEEEAPPSPAEDEHNQNGNLD | |
0.1 mg | |
Apoptosis, Signal Transduction | |
5153 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG3 κ |
Western Blot, ELISA | |
4F9 | |
Western Blot, ELISA | |
AAH32226 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |