Learn More
3M™ PELTOR™ Optime™ I Earmuffs
PELTOR™ Optime™ I Ear Muffs are very lightweight, providing high wearer comfort. Ideal for protection against noise arising from a wide range of applications in the workplace and leisure activities. Offer versatile protection for both short and long duration tasks.
216.00 DKK - 5595.00 DKK
Specifications
Certifications/Compliance | CE Approved, EN 352-3: 2002 |
---|---|
Product Type | Ear Muffs |
Product Code | Brand | Model | Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Style | Price | Quantity & Availability | |||||
15709697
|
3M™
7000038206 |
H510P3E-469-GB | Hard Hat | N/A | ||||||
15709907
|
3M™
7000107744 |
H510P3A-405- GU | Hard Hat | N/A | ||||||
15719707
|
3M™
7000039617 |
H510P3E-405-GU | Hard Hat |
4815.00 DKK
Pack of 20 |
Estimated Shipment: 12-08-2025
Log in to see stock. |
|||||
15719857
|
3M™
7000103737 |
H510A-470-GB | Over-the-head |
216.00 DKK
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||
15719907
|
3M™
7000107745 |
H510P3B-405-GU | Hard Hat | N/A | ||||||
17281806
|
3M™
7000039618 |
H510B-403-GU | Neckband |
2385.00 DKK
Pack of 10 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
15729857
|
3M™
7000103738 |
H510F-459-GB | Over-the-head | N/A | ||||||
15729907
|
3M™
7000107747 |
H510P3EA-405-GU | Hard Hat | N/A | ||||||
15739907
|
3M™
7000107750 |
H510P3G-405-GU | Hard Hat | N/A | ||||||
15749907
|
3M™
7000107751 |
H510P3H-405-GU | Hard Hat | N/A | ||||||
15759907
|
3M™
7000107752 |
H510P3K-405-GU | Hard Hat | N/A | ||||||
15769907
|
3M™
7000107753 |
H510P3A-405-GU | Hard Hat | N/A | ||||||
15789727
|
3M™
7000043332 |
H510P3AF-405-GU | Hard Hat | N/A | ||||||
15799687
|
3M™
7000038205 |
H510F-404-GU | Over-the-head |
2870.00 DKK
Pack of 10 |
Estimated Shipment: 12-08-2025
Log in to see stock. |
|||||
Description
- Low-profile design which minimizes obtrusiveness
- Large space inside cup helps reduce moisture and heat build-up
- Easy-to-replace cushions and inserts helps keep them hygienically clean
- Soft, wide cushions help reduce pressure around the ears and improve comfort and wearability
Specifications
CE Approved, EN 352-3: 2002 | |
Ear Muffs |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
3M™ PELTOR™ Optime™ I Earmuffs