Learn More
3M™ PELTOR™ ProTac™ III Slim Headset Headband
Click to view available options
Slim Headset Headband Earmuff
Description
- Built-in level-dependent function allows wearer to hear ambient sounds, such as conversations, machine sounds and warning signals
- 3.5mm listen-only stereo input (limited to 82dB) for connection to external devices (e.g. mobile phone, radio, two-way radio and iPod)
- Voice-guided menu system avoids need to remove headset for setting changes
- Auto power off turns headset off after 4 hours of non-use, to save energy
- Low-battery warning
- To reduce corrosion caused by sweat, electronics are located in outer part of cup
Specifications
Specifications
Color | Black |
Product Type | Ear Muff with headset headband |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ PELTOR™ ProTac™ III Slim Headset Headband >
Spot an opportunity for improvement?Share a Content Correction