Learn More
Pemirolast potassium, MedChemExpress
MedChemExpress Pemirolast potassium (TWT-8152) is a histamine H1 antagonist and mast cell stabilizer that acts as an antiallergic agent.

634.49 DKK - 1712.11 DKK
Specifications
Chemical Name or Material | Pemirolast potassium |
---|---|
Physical Form | Solid |
CAS | 100299-08-9 |
Color | Yellowish Green |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16494387
![]() |
Medchem Express
HY-B0538A |
50 mg |
1148.12 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16488356
![]() |
Medchem Express
HY-B0538A |
100 mg |
1712.11 DKK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16478518
![]() |
Medchem Express
HY-B0538A |
10 mg |
634.49 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pemirolast potassium (TWT-8152) is a histamine H1 antagonist and mast cell stabilizer that acts as an antiallergic agent.Specifications
Pemirolast potassium | |
100299-08-9 | |
Research | |
COVID-19-immunoregulation | |
H2O : 50 mg/mL (187.76 mM; Need ultrasonic) | |
266.3 | |
99.93% |
Solid | |
Yellowish Green | |
C10H7KN6O | |
TWT-8152 BMY 26517 | |
O=C1C(C2=N[N-]N=N2)=CN=C3N1C=CC=C3C.[K+] | |
266.3 | |
Research |
Safety and Handling
H302
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.