Learn More
Thermo Scientific Chemicals Peptone, from meat (pork gelatine)
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
â—Ź Peptone, from meat, is a growth-promoting nutrient used in bacteria and fungi cultures
Applications
â—Ź It is commonly used in animal cell culture.
â—Ź Peptone is used in mass scale organism cultivation, and fermentation processes
â—Ź Peptone, from meat, is used as a supplement to in vitro culture media to help to increase cell number, specific monoclonal antibody production, and maintain long-term cultures
â—Ź This compound has been used as a component in Ax-2 axenic medium to induce reproduction of Dictyostelium discoideum

Specifications
Specifications
Quantity | 1 kg |
Solubility Information | Solubility aq. soln.: soluble. |
Chemical Name or Material | Peptone |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
A0471052 | Certificate of Analysis | 17/05/2025 | 458930050,458932500,458930010 |
A0463301 | Certificate of Analysis | 21/06/2024 | 458930050,458932500,458930010 |
A0451630 | Certificate of Analysis | 02/05/2023 | 458930050,458932500,458930010 |
A0421191 | Certificate of Analysis | 07/02/2022 | 458930050,458932500,458930010 |
A0435595 | Certificate of Analysis | 06/11/2021 |
Safety and Handling
- Peptone from meat (pork gelatine)
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.