Learn More
Perchloromethyl mercaptan, 97%, Thermo Scientific Chemicals
395.91 DKK - 1222.22 DKK
Chemical Identifiers
CAS | 594-42-3 |
---|---|
Molecular Formula | CCl4S |
Molecular Weight (g/mol) | 185.87 |
MDL Number | MFCD00037054 |
InChI Key | RYFZYYUIAZYQLC-UHFFFAOYSA-N |
Synonym | perchloromethyl mercaptan, perchloromethylmercaptan, clairsit, trichloromethanesulfenyl chloride, methanesulfenyl chloride, trichloro, perchloromethanethiol, perchlormethylmerkaptan, trichloromethane sulfenyl chloride, trichloromethylsulfenyl chloride, trichloromethanesulphenyl chloride |
PubChem CID | 11666 |
IUPAC Name | trichloromethyl thiohypochlorite |
SMILES | ClSC(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10153130
|
Thermo Scientific Acros
130030050 |
5 g | Glass bottle |
395.91 DKK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10537621
|
Thermo Scientific Acros
130030250 |
25 g | Glass bottle |
1222.22 DKK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
594-42-3 | |
185.87 | |
RYFZYYUIAZYQLC-UHFFFAOYSA-N | |
11666 | |
ClSC(Cl)(Cl)Cl |
CCl4S | |
MFCD00037054 | |
perchloromethyl mercaptan, perchloromethylmercaptan, clairsit, trichloromethanesulfenyl chloride, methanesulfenyl chloride, trichloro, perchloromethanethiol, perchlormethylmerkaptan, trichloromethane sulfenyl chloride, trichloromethylsulfenyl chloride, trichloromethanesulphenyl chloride | |
trichloromethyl thiohypochlorite |
Specifications
594-42-3 | |
1.7000g/mL | |
Authentic | |
CCl4S | |
1.5370 to 1.5410 | |
03,135 | |
perchloromethyl mercaptan, perchloromethylmercaptan, clairsit, trichloromethanesulfenyl chloride, methanesulfenyl chloride, trichloro, perchloromethanethiol, perchlormethylmerkaptan, trichloromethane sulfenyl chloride, trichloromethylsulfenyl chloride, trichloromethanesulphenyl chloride | |
ClSC(Cl)(Cl)Cl | |
185.87 | |
185.88 | |
Liquid |
Yellow | |
146.0°C to 148.0°C | |
97% | |
CCl3SCl | |
MFCD00037054 | |
1.7 | |
RYFZYYUIAZYQLC-UHFFFAOYSA-N | |
trichloromethyl thiohypochlorite | |
11666 | |
97% | |
Perchloromethyl mercaptan, 97% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Fatal if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-840-4
missing translation for 'rtecsNumber' : PB0370000
missing translation for 'tsca' : TSCA
- Perchloromethyl mercaptan
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H330-Fatal if inhaled.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
RUO – Research Use Only