missing translation for 'onlineSavingsMsg'
Learn More
Peretinoin, MedChemExpress Product Code.: 16464196

Peretinoin, MedChemExpress

Product Code. p-200100744
100 mg, 100mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16464196

Brand: Medchem Express HY100008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Peretinoin is an oral acyclic retinoid with a vitamin A-like structure that targets retinoid nuclear receptors such as retinoid X receptor (RXR) and retinoic acid receptor (RAR).

Peretinoin reduces the mRNA level of sphingosine kinase 1 (SPHK1) in vitro by downregulating a transcription factor, Sp1. Peretinoin prevents the progression of non-alcoholic steatohepatitis (NASH) and the development of hepatocellular carcinoma (HCC) through activating the autophagy pathway by increased Atg16L1 expression. Peretinoin inhibits HCV RNA amplification and virus release by altering lipid metabolism with a EC50 of 9 ÎĽM.

Specifications

Chemical Name or Material Peretinoin
CAS 81485-25-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H30O2
Quantity 100 mg
Synonym NIK333
Solubility Information DMSO : 50 mg/mL (165.32 mM; Need ultrasonic)
SMILES C/C(C)=C/CC/C(C)=C/CC/C(C)=C/C=C/C(C)=C/C(O)=O
Molecular Weight (g/mol) 302.45
Formula Weight 302.45
Percent Purity 95.66%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Peretinoin, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.