missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Perfect Poly™ Finger Gloves

Product Code. 11871741
Click to view available options
Size:
6
7
8
This item is not returnable. View return policy
This item is not returnable. View return policy

White polyamide knitted gloves with polyurethane-coated fingertips provide dexterity, a snug fit, and good grip for precision work. Honeywell Safety Products™ Perfect Poly™ Finger Gloves are knitted for breathability and comfort and offer excellent abrasion and tear resistance.

  • White, non-pilling polyamide knitted glove suitable for precision work.
  • Suitable for handling fine products in a dry environment.
  • Excellent abrasion and tear resistance.
  • Silicone-free and suitable for painting environments.
  • White polyurethane coating on fingertips for good grip.
  • Tight knit provides comfort, support, dexterity, touch and limits dirt penetration.
  • Ventilated back and palm optimize breathability.
  • Elastic wrist.
  • White color enhances the glove's hygiene and cleanliness.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY I Minor risks, EN 420-2003 Basic standard, EC Category PPE 1, EC Certificate Number Autocertification
Classification Dexterity: 5
Coated Area Fingers
Description Gauge 15
Coating Material Polyurethane
Color White
Material Polyurethane, Polyamide
Product Type Gloves
Size 6
For Use With (Application) Automotive, Aviation and Utility industries
Includes Elasticized wrist
Quantity 10Pair/PK
Length (Metric) 23 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'