missing translation for 'onlineSavingsMsg'
Learn More
Petroleum Ether 40-60°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™ Product Code.: 15374528
compliance-icons compliance-icons compliance-icons compliance-icons

Petroleum Ether 40-60°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™

Product Code. 15374528
2.5 L, Amber Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Fluorinated HDPE Bottle
Metal Can
Plastic-coated Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15374528

Brand: Fisher Chemical P/1760/17X

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

C6H14, CAS Number-64742-49-0, 1,1-dimethylbutane, isohexane, 2-methylpentan, dimethylpropylmethane, 2-methyl-pentane, 2-methyl pentane, unii-49ib0u6mld, pentane, 2-methyl, methyl pentane, iso-hexane, 2.5L, 40 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
PubChem CID 7892
SMILES CCCC(C)C

Specifications

Color Colorless
Boiling Point 40°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 38 kPa at 20°C
Viscosity 0.45 mm2/s at 25°C (ASTM D445)
Synonym Isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, Dimethylpropylmethane
IUPAC Name Petroleum ether

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2525684 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog NumberP/1760/17Z,P/1760/27,P/1760/15,P/1760/17,P/1760/17X,P/1760/25,P/1760/25SS,P/1760/PB17,P/1760/21
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether, boiling range 40-60°C
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Hydrocarbons, C6, isoalkanes < 5% n-hexane (Iso-Hexane)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Petroleum Ether 40-60°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.